siRNA Details
| Gene Symbol: Gm340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145489.1 | - | 2 | 1149 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm340| Gene Symbol: Gm340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145489.1 | - | 2 | 1149 | ||