siRNA Details
| Gene Symbol: Cfap58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163267.1 | NP_001156739.1 | 3 | 540 | ||
| GenBank mRNA | BC057135.1 | - | 3 | 539 | ||
| BC147530.1 | - | 3 | 529 | 9056521 | ||
| BC147540.1 | - | 3 | 529 | 9056531 | ||
| BC150978.1 | - | 3 | 530 | 9087891 | ||
| BC150979.1 | - | 3 | 529 | 9087892 | ||

Cfap58
Genomic Map