siRNA Details
| Gene Symbol: Kif14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287179.2 | NP_001274108.1 | 20 | 3594 | ||
| GenBank mRNA | AB731728.1 | - | 20 | 3484 | ||
| BC054123.1 | - | 8 | 882 | |||
| BC158132.1 | - | 20 | 3507 | 9007399 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kif14| Gene Symbol: Kif14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287179.2 | NP_001274108.1 | 20 | 3594 | ||
| GenBank mRNA | AB731728.1 | - | 20 | 3484 | ||
| BC054123.1 | - | 8 | 882 | |||
| BC158132.1 | - | 20 | 3507 | 9007399 | ||