siRNA Details
| Gene Symbol: Rc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024952.2 | NP_001020123.1 | 5 | 1087 | ||
| GenBank mRNA | AK139648.1 | - | 5 | 1059 | ||
| AK173335.1 | - | 5 | 1033 | |||
| AY948287.1 | - | 5 | 1028 | |||
| BC138663.1 | - | 4 | 811 | 8861684 | ||

Rc3h1
Genomic Map