siRNA Details
| Gene Symbol: Gm996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005424.2 | NP_001005424.2 | 1 | 1046 | ||
| GenBank mRNA | BC065698.1 | - | 2 | 947 | 6825178 | |
| BY097517.1 | - | 1 | 308 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm996| Gene Symbol: Gm996 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005424.2 | NP_001005424.2 | 1 | 1046 | ||
| GenBank mRNA | BC065698.1 | - | 2 | 947 | 6825178 | |
| BY097517.1 | - | 1 | 308 | |||