siRNA Details
| Gene Symbol: Cacfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243239.1 | NP_001230168.1 | 3 | 330 | ||
| NM_001243240.1 | NP_001230169.1 | 3 | 330 | |||
| NM_029862.4 | NP_084138.1 | 3 | 330 | |||
| GenBank mRNA | AK020041.1 | - | 3 | 314 | ||
| AK034046.1 | - | 3 | 314 | |||
| AK038728.1 | - | 3 | 276 | |||
| AK039867.1 | - | 3 | 307 | |||
| AK040379.1 | - | 3 | 237 | |||
| AK043562.1 | - | 3 | 276 | |||
| AK052562.1 | - | 3 | 291 | |||
| AK169470.1 | - | 4 | 306 | |||

Cacfd1
Genomic Map