siRNA Details
| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114079.2 | NP_001107551.1 | 2 | 481 | ||
| GenBank mRNA | BC158030.1 | - | 2 | 481 | 9007333 | |
| EU669908.1 | - | 2 | 224 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc1l| Gene Symbol: Pabpc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114079.2 | NP_001107551.1 | 2 | 481 | ||
| GenBank mRNA | BC158030.1 | - | 2 | 481 | 9007333 | |
| EU669908.1 | - | 2 | 224 | |||