siRNA Details
| Gene Symbol: Zfp663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005425.1 | NP_001005425.1 | 4 | 476 | ||
| GenBank mRNA | AK141109.1 | - | 5 | 616 | ||
| BC067000.1 | - | 4 | 476 | 5705933 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp663| Gene Symbol: Zfp663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005425.1 | NP_001005425.1 | 4 | 476 | ||
| GenBank mRNA | AK141109.1 | - | 5 | 616 | ||
| BC067000.1 | - | 4 | 476 | 5705933 | ||