siRNA Details
| Gene Symbol: Crnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081200.1 | NP_001074669.1 | 3 | 273 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB618328.1 | - | 273 | Chr. - - [-] on Build | ||

Crnn
Genomic Map