siRNA Details
| Gene Symbol: Nr1h5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198658.2 | NP_941060.2 | 10 | 1323 | ||
| NM_201619.2 | NP_963913.2 | 10 | 1299 | |||
| NM_201622.2 | NP_963916.2 | 10 | 1149 | |||
| GenBank mRNA | AY094586.1 | - | 10 | 1323 | ||
| AY094587.1 | - | 9 | 1218 | |||
| AY094588.1 | - | 9 | 1194 | |||
| AY094589.1 | - | 9 | 1175 | |||
| AY094590.1 | - | 9 | 1151 | |||
| AY094591.1 | - | 9 | 1044 | |||
| BC119524.1 | - | 9 | 1218 | 40056073 | ||
| BC119525.1 | - | 9 | 1194 | 40056074 | ||

Nr1h5
Genomic Map