siRNA Details
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081281.1 | NP_001074750.1 | 2 | 465 | ||
GenBank mRNA | AK052918.1 | - | 2 | 466 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trim55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081281.1 | NP_001074750.1 | 2 | 465 | ||
GenBank mRNA | AK052918.1 | - | 2 | 466 |