siRNA Details
| Gene Symbol: Dpy19l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081201.1 | NP_001074670.1 | 6 | 713 | ||
| GenBank mRNA | AK144417.1 | - | 6 | 665 | ||
| AK165377.1 | - | 6 | 658 | |||
| AK172656.1 | - | 6 | 721 | |||
| BC147500.1 | - | 6 | 669 | 9056363 | ||
| BC157932.1 | - | 6 | 669 | 9088105 | ||
| BC157988.1 | - | 6 | 669 | 9088143 | ||

Dpy19l4
Genomic Map