siRNA Details
| Gene Symbol: Dpy19l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081201.1 | NP_001074670.1 | 18, 19 | 2124 | ||
| GenBank mRNA | AK144417.1 | - | 18, 19 | 2076 | ||
| BC147500.1 | - | 18, 19 | 2080 | 9056363 | ||
| BC157932.1 | - | 18, 19 | 2080 | 9088105 | ||
| BC157988.1 | - | 18, 19 | 2080 | 9088143 | ||

Dpy19l4
Genomic Map