siRNA Details
| Gene Symbol: AI427809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144542.1 | - | 2 | 1058 | ||
| AK171862.1 | - | 2 | 1075 | |||
| BC147516.1 | - | 1 | 215 | 9056508 | ||
| BC147547.1 | - | 1 | 215 | 9056538 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI427809| Gene Symbol: AI427809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144542.1 | - | 2 | 1058 | ||
| AK171862.1 | - | 2 | 1075 | |||
| BC147516.1 | - | 1 | 215 | 9056508 | ||
| BC147547.1 | - | 1 | 215 | 9056538 | ||