siRNA Details
| Gene Symbol: Mup20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012323.1 | NP_001012323.1 | 7 | 769 | ||
| GenBank mRNA | BC089613.1 | - | 7 | 769 | 6746470 | |
| BC092096.1 | - | 7 | 795 | 4161847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mup20| Gene Symbol: Mup20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012323.1 | NP_001012323.1 | 7 | 769 | ||
| GenBank mRNA | BC089613.1 | - | 7 | 769 | 6746470 | |
| BC092096.1 | - | 7 | 795 | 4161847 | ||