siRNA Details
| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033773.4 | NP_001028945.2 | 4 | 570 | ||
| GenBank mRNA | AK133379.1 | - | 3 | 489 | ||
| BB616918.1 | - | 3 | 489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2u| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033773.4 | NP_001028945.2 | 4 | 570 | ||
| GenBank mRNA | AK133379.1 | - | 3 | 489 | ||
| BB616918.1 | - | 3 | 489 | |||