siRNA Details
| Gene Symbol: Oog2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198661.3 | NP_941063.1 | 3 | 384 | ||
| GenBank mRNA | AK139660.1 | - | 2 | 856 | ||
| AY573562.1 | - | 3 | 384 | |||
| BC052839.1 | - | 3 | 384 | 30050073 | ||
| Gene Symbol: Gm13084 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005371.3 | NP_001005371.2 | 2 | 322 | ||
| GenBank mRNA | AK145369.1 | - | 2 | 319 | ||
| BC080785.1 | - | 2 | 320 | 30481291 | ||
| CJ082474.1 | - | 2 | 323 | |||

Oog2
Genomic Map