siRNA Details
| Gene Symbol: C87499 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198663.3 | NP_941065.2 | 3 | 658 | ||
| GenBank mRNA | AK135883.1 | - | 3 | 658 | ||
| AK163216.1 | - | 2 | 349 | |||
| BC053719.1 | - | 3 | 659 | 30053512 | ||
| CJ083078.1 | - | 2 | 348 | |||

C87499
Genomic Map