siRNA Details
| Gene Symbol: Adgrf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014394.2 | NP_001014416.2 | 11, 12 | 2847 | ||
| GenBank mRNA | AY701869.1 | - | 11, 12 | 2847 | ||
More Information
Other Species Targeted
Set Membership: |
|

Adgrf3
Genomic Map