siRNA Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 12 | 1709 | ||
| GenBank mRNA | AK076392.1 | - | 5 | 575 | ||
| AK137244.1 | - | 13 | 1936 | |||
| AK172988.1 | - | 12 | 1620 | |||
| BC062951.1 | - | 12 | 1710 | 6852711 | ||

Cep135
Genomic Map