siRNA Details
| Gene Symbol: Cep135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199032.2 | NP_950197.1 | 10 | 1461 | ||
| GenBank mRNA | AK076392.1 | - | 3 | 327 | ||
| AK137244.1 | - | 11 | 1688 | |||
| AK172988.1 | - | 10 | 1372 | |||
| BC062951.1 | - | 10 | 1462 | 6852711 | ||

Cep135
Genomic Map