siRNA Details
| Gene Symbol: Lrcol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033459.3 | NP_001028631.1 | 1 | 169 | ||
| GenBank mRNA | AK141249.1 | - | 1 | 170 | ||
| BC147447.1 | - | 1 | 107 | 9056089 | ||
| BC147469.1 | - | 1 | 107 | 9056111 | ||
| BC171993.1 | - | 1 | 107 | |||
| JQ258939.1 | - | 3 | 314 | |||
| JQ258940.1 | - | 4 | 377 | |||
| JQ258941.1 | - | 4 | 377 | |||

Lrcol1
Genomic Map