siRNA Details
| Gene Symbol: Lrcol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033459.3 | NP_001028631.1 | 2 | 282 | ||
| GenBank mRNA | AK141249.1 | - | 2 | 283 | ||
| BC147447.1 | - | 2 | 220 | 9056089 | ||
| BC147469.1 | - | 2 | 220 | 9056111 | ||
| BC171993.1 | - | 2 | 198 | |||
| JQ258939.1 | - | 4 | 427 | |||
| JQ258940.1 | - | 5 | 490 | |||
| JQ258941.1 | - | 4 | 687 | |||

Lrcol1
Genomic Map