siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142642.1 | NP_001136114.1 | 11, 12 | 1922 | ||
| NM_028596.2 | NP_082872.2 | 5, 6 | 1038 | |||
| GenBank mRNA | AK031967.1 | - | 5, 6 | 1008 | ||
| AK211844.1 | - | 2, 3 | 185 | |||
| BC059932.1 | - | 9, 10 | 2708 | |||
| BC151196.1 | - | 6, 7 | 931 | 9088084 | ||
| BU614458.1 | - | 2, 3 | 230 | |||

Fbrsl1
Genomic Map