siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142642.1 | NP_001136114.1 | 18, 19 | 2594 | ||
| NM_028596.2 | NP_082872.2 | 11, 12 | 1638 | |||
| GenBank mRNA | AK031967.1 | - | 11, 12 | 1608 | ||
| AK129388.1 | - | 7, 8 | 3150 | |||
| BC059932.1 | - | 16, 17 | 3380 | |||
| BC069994.1 | - | 6, 7 | 595 | |||
| BC151196.1 | - | 13, 14 | 1603 | 9088084 | ||

Fbrsl1
Genomic Map