siRNA Details
| Gene Symbol: Fbrsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142642.1 | NP_001136114.1 | 10 | 1696 | ||
| NM_028596.2 | NP_082872.2 | 4 | 812 | |||
| GenBank mRNA | AK031967.1 | - | 4 | 782 | ||
| BC059932.1 | - | 9 | 1852 | |||
| BC151196.1 | - | 5 | 693 | 9088084 | ||
| CX204847.1 | - | 3 | 228 | |||

Fbrsl1
Genomic Map