siRNA Details
| Gene Symbol: Zcwpw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005426.2 | NP_001005426.2 | 7 | 854 | ||
| GenBank mRNA | BC071186.1 | - | 7 | 781 | 30107853 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcwpw1| Gene Symbol: Zcwpw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005426.2 | NP_001005426.2 | 7 | 854 | ||
| GenBank mRNA | BC071186.1 | - | 7 | 781 | 30107853 | |