siRNA Details
Gene Symbol: Zcwpw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005426.2 | NP_001005426.2 | 12 | 1378 | ||
GenBank mRNA | BC071186.1 | - | 12 | 1305 | 30107853 | |
CF982070.1 | - | 2 | 99 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zcwpw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005426.2 | NP_001005426.2 | 12 | 1378 | ||
GenBank mRNA | BC071186.1 | - | 12 | 1305 | 30107853 | |
CF982070.1 | - | 2 | 99 |