siRNA Details
| Gene Symbol: Wdr95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029440.3 | NP_083716.2 | 15 | 1273 | ||
| GenBank mRNA | AK015309.1 | - | 13 | 1377 | ||
| AK087389.1 | - | 15 | 1273 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr95| Gene Symbol: Wdr95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029440.3 | NP_083716.2 | 15 | 1273 | ||
| GenBank mRNA | AK015309.1 | - | 13 | 1377 | ||
| AK087389.1 | - | 15 | 1273 | |||