siRNA Details
| Gene Symbol: N4bp2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201369.3 | NP_958757.2 | 2 | 684 | ||
| GenBank mRNA | BC037393.1 | - | 2 | 649 | ||
| CB231597.1 | - | 1 | 109 | |||
| CF724618.1 | - | 2 | 591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

N4bp2l2| Gene Symbol: N4bp2l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201369.3 | NP_958757.2 | 2 | 684 | ||
| GenBank mRNA | BC037393.1 | - | 2 | 649 | ||
| CB231597.1 | - | 1 | 109 | |||
| CF724618.1 | - | 2 | 591 | |||