siRNA Details
Gene Symbol: N4bp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201369.3 | NP_958757.2 | 2 | 1038 | ||
GenBank mRNA | BC037393.1 | - | 2 | 1003 | ||
CB231597.1 | - | 1 | 463 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: N4bp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201369.3 | NP_958757.2 | 2 | 1038 | ||
GenBank mRNA | BC037393.1 | - | 2 | 1003 | ||
CB231597.1 | - | 1 | 463 |