siRNA Details
| Gene Symbol: Gm9758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198666.3 | NP_941068.1 | 4 | 554 | ||
| GenBank mRNA | BC048454.1 | - | 4 | 554 | 6741646 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm9758| Gene Symbol: Gm9758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198666.3 | NP_941068.1 | 4 | 554 | ||
| GenBank mRNA | BC048454.1 | - | 4 | 554 | 6741646 | |