siRNA Details
| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122730.2 | NP_001116202.2 | 23 | 6815 | ||
| GenBank mRNA | BC094365.1 | - | 4 | 344 | ||
| CB244319.1 | - | 6 | 570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc18| Gene Symbol: Tnrc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122730.2 | NP_001116202.2 | 23 | 6815 | ||
| GenBank mRNA | BC094365.1 | - | 4 | 344 | ||
| CB244319.1 | - | 6 | 570 | |||