siRNA Details
| Gene Symbol: Tsen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199033.1 | NP_950198.1 | 8 | 1071 | ||
| GenBank mRNA | AK134697.1 | - | 8 | 1067 | ||
| BC061473.1 | - | 8 | 1071 | 6414790 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tsen2| Gene Symbol: Tsen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199033.1 | NP_950198.1 | 8 | 1071 | ||
| GenBank mRNA | AK134697.1 | - | 8 | 1067 | ||
| BC061473.1 | - | 8 | 1071 | 6414790 | ||