siRNA Details
Gene Symbol: Cracr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033464.3 | NP_001028636.1 | 6 | 819 | ||
GenBank mRNA | AK143832.1 | - | 6 | 820 | ||
BC150869.1 | - | 6 | 897 | 9087780 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cracr2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033464.3 | NP_001028636.1 | 6 | 819 | ||
GenBank mRNA | AK143832.1 | - | 6 | 820 | ||
BC150869.1 | - | 6 | 897 | 9087780 |