siRNA Details
Gene Symbol: 4922502D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199034.3 | NP_950199.1 | 4 | 661 | ||
GenBank mRNA | AK076635.1 | - | 4 | 634 | ||
AK132711.1 | - | 4 | 646 | |||
BC089479.1 | - | 4 | 661 | 6704696 | ||
GU553093.1 | - | 4 | 444 |
