siRNA Details
| Gene Symbol: 4922502D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199034.3 | NP_950199.1 | 3, 4 | 555 | ||
| GenBank mRNA | AK076635.1 | - | 3, 4 | 528 | ||
| AK132711.1 | - | 3, 4 | 540 | |||
| BC089479.1 | - | 3, 4 | 555 | 6704696 | ||
| GU553093.1 | - | 3, 4 | 338 | |||

4922502D21Rik
Genomic Map