siRNA Details
| Gene Symbol: Alg8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199035.2 | NP_950200.2 | 3 | 252 | ||
| GenBank mRNA | AK141341.1 | - | 3 | 246 | ||
| AK166755.1 | - | 3 | 247 | |||
| AK166897.1 | - | 3 | 253 | |||
| BC061244.1 | - | 3 | 241 | 30246181 | ||

Alg8
Genomic Map