siRNA Details
| Gene Symbol: Ppapdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080963.1 | NP_001074432.1 | 2 | 90 | ||
| GenBank mRNA | AK081292.1 | - | 2 | 154 | ||
| BC120497.1 | - | 2 | 90 | 40129420 | ||
| BC144735.1 | - | 2 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppapdc1a| Gene Symbol: Ppapdc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080963.1 | NP_001074432.1 | 2 | 90 | ||
| GenBank mRNA | AK081292.1 | - | 2 | 154 | ||
| BC120497.1 | - | 2 | 90 | 40129420 | ||
| BC144735.1 | - | 2 | 90 | |||