siRNA Details
| Gene Symbol: 6430531B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033465.2 | NP_001028637.1 | 4 | 615 | ||
| GenBank mRNA | AK131789.1 | - | 4 | 616 | ||
| AK164921.1 | - | 4 | 585 | |||
| BC025923.1 | - | 3 | 951 | |||
| BC145460.1 | - | 4 | 579 | |||
| BC145461.1 | - | 4 | 579 | |||
| BC145730.1 | - | 4 | 579 | 40131033 | ||
| BC171930.1 | - | 4 | 579 | |||

6430531B16Rik
Genomic Map