siRNA Details
| Gene Symbol: Usp17lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201409.2 | NP_958811.2 | 2 | 1203 | ||
| GenBank mRNA | AY257250.1 | - | 2 | 1200 | ||
| BC137942.1 | - | 2 | 1299 | 8860961 | ||
| BC145058.1 | - | 2 | 1299 | 9053606 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp17lb| Gene Symbol: Usp17lb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201409.2 | NP_958811.2 | 2 | 1203 | ||
| GenBank mRNA | AY257250.1 | - | 2 | 1200 | ||
| BC137942.1 | - | 2 | 1299 | 8860961 | ||
| BC145058.1 | - | 2 | 1299 | 9053606 | ||