siRNA Details
| Gene Symbol: Cnep1r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029074.3 | NP_083350.2 | 4 | 293 | ||
| GenBank mRNA | AK017198.1 | - | 4 | 254 | ||
| AK146578.1 | - | 4 | 284 | |||
| AK217813.1 | - | 1 | 23 | |||
| BC018498.1 | - | 3 | 191 | |||
| BC094385.1 | - | 4 | 252 | |||
| BC118981.1 | - | 4 | 254 | |||
| BC148291.2 | - | 4 | 254 | 40103934 | ||

Cnep1r1
Genomic Map