siRNA Details
| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033469.2 | NP_001028641.1 | 5 | 682 | ||
| GenBank mRNA | AK139562.1 | - | 5 | 682 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxr1| Gene Symbol: Foxr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033469.2 | NP_001028641.1 | 5 | 682 | ||
| GenBank mRNA | AK139562.1 | - | 5 | 682 | ||