siRNA Details
| Gene Symbol: Ccdc33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166282.1 | NP_001159754.1 | 7 | 1302 | ||
| NM_029212.3 | NP_083488.1 | 6 | 879 | |||
| GenBank mRNA | AB048860.1 | - | 6 | 953 | ||
| AK145264.1 | - | 7 | 1304 | |||
| BC116291.1 | - | 6 | 879 | 40094912 | ||
| BC116292.1 | - | 6 | 879 | 40094918 | ||

Ccdc33
Genomic Map