siRNA Details
| Gene Symbol: Fbxw22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014395.2 | NP_001014417.1 | 10 | 1379 | ||
| GenBank mRNA | AK135808.1 | - | 8 | 1096 | ||
| AK162068.1 | - | 6 | 887 | |||
| BC084593.1 | - | 10 | 1383 | 30477093 | ||
| Gene Symbol: Fbxw14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135771.1 | - | 1379 | |||

Fbxw22
Genomic Map