siRNA Details
| Gene Symbol: Fbxw22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014395.2 | NP_001014417.1 | 9, 10 | 1334 | ||
| GenBank mRNA | AK135808.1 | - | 7, 8 | 1051 | ||
| AK162068.1 | - | 5, 6 | 842 | |||
| BC084593.1 | - | 9, 10 | 1338 | 30477093 | ||

Fbxw22
Genomic Map