siRNA Details
| Gene Symbol: Gm1140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126317.1 | NP_001119789.1 | 4 | 234 | ||
| GenBank mRNA | BG088488.2 | - | 4 | 233 | ||
| Gene Symbol: Gm14692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163195.1 | NP_001156667.1 | 4 | 234 | ||

Gm1140
Genomic Map