siRNA Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164683.1 | NP_001158155.1 | 2 | 254 | ||
| NM_001164684.1 | NP_001158156.1 | 2 | 223 | |||
| NM_001164686.1 | NP_001158158.1 | 2 | 204 | |||
| GenBank mRNA | AK012546.1 | - | 2 | 215 | ||
| AK018228.1 | - | 2 | 224 | |||
| AK154711.1 | - | 2 | 258 | |||
| BC058428.1 | - | 2 | 165 | |||
| BY288280.1 | - | 2 | 254 | |||
| CA526769.1 | - | 2 | 205 | |||
| CK624690.1 | - | 2 | 175 | |||

Tmem29
Genomic Map