siRNA Details
| Gene Symbol: Cdkl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024624.2 | NP_001019795.1 | 5 | 494 | ||
| GenBank mRNA | AK052380.1 | - | 5 | 491 | ||
| AK139220.1 | - | 5 | 466 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdkl5| Gene Symbol: Cdkl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024624.2 | NP_001019795.1 | 5 | 494 | ||
| GenBank mRNA | AK052380.1 | - | 5 | 491 | ||
| AK139220.1 | - | 5 | 466 | |||