siRNA Details
| Gene Symbol: Gm5169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040669.1 | NP_001035759.1 | 8 | 633 | ||
| GenBank mRNA | BC087929.1 | - | 8 | 633 | 6742946 | |
| Gene Symbol: Gm2012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104946.2 | NP_001098416.1 | 8 | 636 | ||

Gm5169
Genomic Map